2,4-Dinitro-5-fluoroaniline ≥98.0%

Supplier: TCI America

Synonyms: 5-Fluoro-2,4-dinitroaniline

D1637-1G
TCD1637-001GEA 68 USD
TCD1637-001G
2,4-Dinitro-5-fluoroaniline ≥98.0%
2,4-Dinitro-5-fluoroaniline

CAS Number: 367-81-7
MDL Number: MFCD00007150
Molecular Formula: C6H4FN3O4
Molecular Weight: 201.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 187


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-Fluoro-2,4-dinitroaniline

Formula: C₆H₄FN₃O₄
MW: 201.11 g/mol
Melting Pt: 187 °C
MDL Number: MFCD00007150
CAS Number: 367-81-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8b3b15873204bcdec08af6f2a73f1e46b9b5509&vid=c8b3b15873204bcdec08af6f2a73f1e46b9b5509&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR