3,3'-Dimethylbiphenyl ≥98.0%

Supplier: TCI America

Synonyms: 3,3'-Bitolyl, 3,3'-Dimethyl-1,1'-biphenyl

D1430-5ML
TCD1430-005MLEA 147.5 USD
TCD1430-005ML
3,3'-Dimethylbiphenyl ≥98.0%
3,3'-Dimethylbiphenyl

CAS Number: 612-75-9
MDL Number: MFCD00008534
Molecular Formula: C14H14
Molecular Weight: 182.27
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 155
Flash Point (°C): 110
Freezing point (°C): 9
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: 3,3'-Bitolyl

Formula: C₁₄H₁₄
MW: 182.27 g/mol
Boiling Pt: 155 °C
Density: 1.00 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008534
CAS Number: 612-75-9
EINECS: 210-319-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6acbba83b0cb66caed2944442f27804eb150043&vid=e6acbba83b0cb66caed2944442f27804eb150043&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR