trans-2,2-Dimethyl-3-heptene ≥99.0%

Supplier: TCI America

Synonyms: (E)-2,2-Dimethyl-3-heptene

D1260-5ML
TCD1260-005MLEA 339 USD
TCD1260-005ML
trans-2,2-Dimethyl-3-heptene ≥99.0%
trans-2,2-Dimethyl-3-heptene

CAS Number: 19550-75-5
MDL Number: MFCD00039961
Molecular Formula: C9H18
Molecular Weight: 126.24
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Flash Point (°C): 19
Specific Gravity (20/20): 0.72

Formula: C₉H₁₈
MW: 126.24 g/mol
Boiling Pt: 127 °C (1013 hPa)
Melting Pt: –85 °C
Density: 0.72 (20 °C)
Flash Pt: 19 °C
MDL Number: MFCD00039961
CAS Number: 19550-75-5
EINECS: 243-156-7

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR