N,N'-Diphenyl-p-phenylenediamine ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: DPPD, N,N′-Diphenyl-1,4-phenylenediamine, N,N'-Diphenyl-1,4-phenylenediamine

D0609-100G
TCD0609-100GEA 149.37 USD
TCD0609-100G
N,N'-Diphenyl-p-phenylenediamine ≥95.0% (by titrimetric analysis)
N,N'-Diphenyl-p-phenylenediamine

CAS Number: 74-31-7
MDL Number: MFCD00003015
Molecular Formula: C18H16N2
Molecular Weight: 260.34
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Boiling point (°C): 225
Melting point (°C): 152
Flash Point (°C): 236

Formula: C₁₈H₁₆N₂
MW: 260.34 g/mol
Boiling Pt: 225 °C
Melting Pt: 152 °C
Density: 1.21 g/cm³ (20 °C)
Flash Pt: 236 °C
MDL Number: MFCD00003015
CAS Number: 74-31-7
EINECS: 200-806-4
Merck Index: 12,03388
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e37197a67a0aecf0b4b2454d329042a116595963&vid=e37197a67a0aecf0b4b2454d329042a116595963&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR