3,4-Dichloroaniline ≥98.0%

Supplier: TCI America
Danger

D0324-25G
TCD0324-25GEA 31 USD
TCD0324-25G
3,4-Dichloroaniline ≥98.0%
3,4-Dichloroaniline

CAS Number: 95-76-1
MDL Number: MFCD00007768
Molecular Formula: C6H5Cl2N
Molecular Weight: 162.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 145
Melting point (°C): 72
Flash Point (°C): 135

Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Boiling Pt: 145 °C
Melting Pt: 72 °C
Density: 1.567 g/cm³ (20 °C)
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00007768
CAS Number: 95-76-1
EINECS: 202-448-4
UN: 3442
ADR: 6.1,II
Merck Index: 13,03079
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8730b733b2cc22a117f526d77770e12061e6449d&vid=9cf202fd9d2ccc6dd9e7f84253d07affe3417e2d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR