(S)-2,4-Diaminobutanoic acid dihydrochloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: L-2,4-Diaminobutanoic acid dihydrochloride, H-Dab-OH 2HCl, L-2,4-Diaminobutyric acid dihydrochloride, (2S)-2,4-Diaminobutanoic acid dihydrochloride, H-Daba-OH·2HCl, (S)-(+)-2,4-Diaminobutyric acid dihydrochloride

D0083-1G
TCD0083-001GEA 63 USD
TCD0083-001G
(S)-2,4-Diaminobutanoic acid dihydrochloride ≥97.0% (by titrimetric analysis)
(S)-2,4-Diaminobutanoic acid dihydrochloride

CAS Number: 1883-09-6
MDL Number: MFCD00064561
Molecular Formula: C4H10N2O2
Molecular Weight: 191.05
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 195
Specific rotation [a]20/D: 14.5 deg (C=3.6, H2O)

Formula: C₄H₁₂Cl₂N₂O₂
MW: 191.06 g/mol
Melting Pt: 195 °C
MDL Number: MFCD00064561
CAS Number: 1883-09-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57561b1ef1cf9e2fe1874903c5d6983b8240de10&vid=57561b1ef1cf9e2fe1874903c5d6983b8240de10&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR