2-Chloro-3-cyano-4-methoxypyridine ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 2-Chloro-4-methoxynicotinonitrile

C3282-1G
TCC3282-1GEA 87 USD
TCC3282-1G
2-Chloro-3-cyano-4-methoxypyridine ≥98.0% (by GC)
2-Chloro-3-cyano-4-methoxypyridine
Formula: C₇H₅ClN₂O
MW: 168.5804 g/mol
MDL Number: MFCD00975444
CAS Number: 98645-43-3
UN: 3439
ADR: 6.1,III
New Product

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 173.0 to 177.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR