4-Chloro-2,6-dimethyl-3-nitropyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

C3146-1G
TCC3146-1GEA 78 USD
TCC3146-1G
4-Chloro-2,6-dimethyl-3-nitropyridine ≥98.0% (by GC)
4-Chloro-2,6-dimethyl-3-nitropyridine
Formula: C₇H₇ClN₂O₂
MDL Number: MFCD08543468
CAS Number: 15513-48-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 75.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR