4-Carboxy-2-chlorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 4-Carboxy-2-chlorobenzeneboronic acid

C2916-1G
TCC2916-1GEA 68 USD
TCC2916-1G
4-Carboxy-2-chlorophenylboronic acid (contains varying amounts of Anhydride)
4-Carboxy-2-chlorophenylboronic acid
Formula: C₇H₆BClO₄
MW: 200.39 g/mol
MDL Number: MFCD07363763
CAS Number: 851335-09-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 98.0 to 110.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR