(4-Chlorophenyl)acetyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 2-(4-Chlorophenyl)acetyl chloride, 4-Chlorobenzeneacetyl chloride

C2313-25G
TCC2313-25GEA 96 USD
TCC2313-25G
(4-Chlorophenyl)acetyl chloride ≥98.0% (by GC, titration analysis)
(4-Chlorophenyl)acetyl chloride

CAS Number: 25026-34-0
MDL Number: MFCD00037111
Molecular Formula: C8H6Cl2O
Molecular Weight: 189.04
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 129
Flash Point (°C): 110
Specific Gravity (20/20): 1.31

Formula: (C₆H₄Cl)CH₂COCl
MW: 189.04 g/mol
Boiling Pt: 129 °C
Density: 1.31 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00037111
CAS Number: 25026-34-0
EINECS: 246-571-1
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d52bc378c840276c8da00c08256412622f7470e9&vid=d52bc378c840276c8da00c08256412622f7470e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR