4-Cyclohexylbenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C1523-5G C1523-25G
TCC1523-005GEA 68 USD
TCC1523-005G TCC1523-025G
4-Cyclohexylbenzoic acid ≥98.0% (by GC, titration analysis)
4-Cyclohexylbenzoic acid

CAS Number: 20029-52-1
MDL Number: MFCD00039459
Molecular Formula: C13H16O2
Molecular Weight: 204.27
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 200

Formula: C₁₃H₁₆O₂
MW: 204.27 g/mol
Melting Pt: 200 °C
MDL Number: MFCD00039459
CAS Number: 20029-52-1
EINECS: 243-472-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6824b38af8e8dd1653a9daccc0ab6042e319f8f&vid=344d9931a92d8d4d7eb87d7c3714dec77ad01536&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR