Crotonic anhydride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

C0420-25G
TCC0420-025GEA 16.28 USD
TCC0420-025G
Crotonic anhydride ≥98.0% (by titrimetric analysis)
Crotonic anhydride

Form: Clear Liquid
Boiling point (°C): 131
Specific Gravity (20/20): 1.04

Formula: C₈H₁₀O₃
MW: 154.17 g/mol
Boiling Pt: 131 °C
Density: 1.04 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009286
CAS Number: 623-68-7
EINECS: 210-807-1
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR