1-Bromodibenzothiophene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Bromodibenzo[b,d]thiophene

B5861-1G
TCB5861-1GEA 176 USD
TCB5861-1G
1-Bromodibenzothiophene ≥98.0% (by GC)
1-Bromodibenzothiophene
Formula: C₁₂H₇BrS
MW: 263.15 g/mol
MDL Number: MFCD18451629
CAS Number: 65642-94-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 82.0 to 86.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR