Benzyl alcohol 99,0

Supplier: TCI America
Warning

Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol

B2378-500G
TCB2378-500GEA 40.41 USD
TCB2378-500G
Benzyl alcohol 99,0
Benzyl alcohol

CAS Number: 100-51-6
MDL Number: MFCD00004599
Molecular Formula: C7H8O
Molecular Weight: 108.14
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 200
Melting point (°C): -15
Flash Point (°C): 100
Specific Gravity (20/20): 1.05

Formula: C₆H₅CH₂OH
MW: 108.14 g/mol
Boiling Pt: 205
Melting Pt: -15.3
Density: 1.05 (20 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a9d53adc3b6b9717df79d414b32ec42d7d3b43f&vid=6a9d53adc3b6b9717df79d414b32ec42d7d3b43f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR