N,O-Bis(trimethylsilyl)hydroxylamine ≥95.0%

Supplier: TCI America

B1825-5G
TCB1825-005GEA 501 USD
TCB1825-005G
N,O-Bis(trimethylsilyl)hydroxylamine ≥95.0%
N,O-Bis(trimethylsilyl)hydroxylamine

CAS Number: 22737-37-7 MDL Number: MFCD00008266 Molecular Formula: C6H19NOSi2 Molecular Weight: 177.39 Purity/Analysis Method: 95.0% (GC) Form: Clear Liquid Flash Point (°C): 29 Specific Gravity (20/20): 0.83 Storage Temperature: 0-10°C

Formula: C₆H₁₉NOSi₂
MW: 177.39 g/mol
Density: 0.83 (20 °C)
Flash Pt: 29 °C
MDL Number: MFCD00008266
CAS Number: 22737-37-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61ac52dd44593cf034f56e993f5669bff2f944f1&vid=61ac52dd44593cf034f56e993f5669bff2f944f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR