(+)-3-Bromocamphor-8-sulfonic acid ammonium salt ≥98.0%

Supplier: TCI America

Synonyms: Ammonium ((1R,3S,4S,7R)-3-bromo-1,7-dimethyl-2-oxobicyclo[2.2.1]heptan-7-yl)methanesulphonate

B1125-5G B1125-25G
TCB1125-005GEA 33 USD
TCB1125-005G TCB1125-025G
(+)-3-Bromocamphor-8-sulfonic acid ammonium salt ≥98.0%
(+)-3-Bromocamphor-8-sulfonic acid ammonium salt

CAS Number: 14575-84-9
MDL Number: MFCD00167438
Molecular Formula: C10H18BrNO4S
Molecular Weight: 328.22
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: 86 deg (C=4, H2O)

Formula: C₁₀H₁₈BrNO₄S
MW: 328.22 g/mol
MDL Number: MFCD00167438
CAS Number: 14575-84-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98c0ab448486c4c2ab1f4826f9c4ac422c581065&vid=625c7c42d2d81ce70aebb4abfab261eb82483afb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR