4,4'-(Hexafluoroisopropylidene)diphenol ≥98.0%

Supplier: TCI America

Synonyms: 4,4′-(Hexafluoroisopropylidene)diphenol, 2,2-Bis(4-hydroxyphenyl)hexafluoropropane, 4,4'-(Perfluoropropane-2,2-diyl)diphenol

B0945-25G B0945-5G B0945-100G
TCB0945-025GEA 49.84 USD
TCB0945-025G TCB0945-005G TCB0945-100G
4,4'-(Hexafluoroisopropylidene)diphenol ≥98.0%
4,4'-(Hexafluoroisopropylidene)diphenol

CAS Number: 1478-61-1
MDL Number: MFCD00000439
Molecular Formula: C15H10F6O2
Molecular Weight: 336.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 161
Flash Point (°C): 205


Product Categories: Materials Chemistry, Monomers, Bisphenol AF type Compounds (for High-Performance Polymer Research), Building Blocks for Liquid Crystals
Other Product Names / Synonyms: Bisphenol AF, Bisphenol A Hexafluoride, Hexafluorobisphenol A, 4,4'-(Hexafluoroisopropylidene)diphenol

Formula: C₁₅H₁₀F₆O₂
MW: 336.24 g/mol
Melting Pt: 161 °C
Flash Pt: 205 °C
MDL Number: MFCD00000439
CAS Number: 1478-61-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d72a5b8ecfdf9913fb0284faeda3c3f0ac7929f&vid=8d72a5b8ecfdf9913fb0284faeda3c3f0ac7929f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR