Lumogallion ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-Chloro-6-(2,4-dihydroxyphenylazo)-1-phenol-2-sulphonic acid, 2,2',4'-Trihydroxy-5-chloroazobenzene-3-sulphonic acid

A5060-1G
TCA5060-001GEA 90 USD
TCA5060-001G
Lumogallion ≥98.0% (by titrimetric analysis)
Lumogallion

[Fluorimetric reagent for Al, Ga and other metals]
CAS Number: 4386-25-8
MDL Number: MFCD00047770
Molecular Formula: C12H9ClN2O6S
Molecular Weight: 344.72
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Analytical Science, Azo Compounds (Chelating Agents)
Other Product Names / Synonyms: 4-Chloro-6-(2,4-dihydroxyphenylazo)-1-phenol-2-sulfonic Acid, 2,2',4'-Trihydroxy-5-chloroazobenzene-3-sulfonic Acid

Formula: C₁₂H₉ClN₂O₆S
MW: 344.72 g/mol
MDL Number: MFCD00047770
CAS Number: 4386-25-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45bfef433c8dffff852c1c2fca47b33bba2a0845&vid=45bfef433c8dffff852c1c2fca47b33bba2a0845&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR