Alosetron hydrochloride ≥98.0% (by HPLC)

Supplier: TCI America
Danger

Synonyms: GR 68755 Hydrochloride, 2,3,4,5-Tetrahydro-5-methyl-2-[(5-methyl-1H-imidazol-4-yl)methyl]-1H-pyrido[4,3-b]indol-1-one hydrochloride, GR 68755C, 5-Methyl-2-[(5-methyl-1H-imidazol-4-yl)methyl]-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indol-1-one Hydrochloride, GR 68755X Hydrochloride, hydrochloride, 5-Methyl-2-[(5-methyl-1H-imidazol-4-yl)methyl]-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indol-1-one hydrochloride, Alosetron Hydrochloride, 5-methyl-2-[(5-methyl-1H-imidazol-4-yl)methyl]-3,4-dihydropyrido[4,3-b]indol-1-one

A3197-100MG
TCA3197-100MGEA 144 USD
TCA3197-100MG
Alosetron hydrochloride ≥98.0% (by HPLC)
Alosetron hydrochloride
Formula: C₁₇H₁₈N₄O·HCl
MW: 330.82 g/mol
Melting Pt: 291 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03453647
CAS Number: 122852-69-1
UN: 2811
ADR: 6.1,III
Merck Index: 14,00308

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR