5-Chloro-2-phenoxyaniline ≥98.0%

Supplier: TCI America

Synonyms: 2-Amino-4-chlorodiphenyl ether, 2-Amino-4-chlorophenyl phenyl ether, 5-Chloro-2-phenoxyaniline, 2-Amino-4-chloro-diphenyl ether

A2222-25G
TCA2222-25GEA 55 USD
TCA2222-25G
5-Chloro-2-phenoxyaniline ≥98.0%
5-Chloro-2-phenoxyaniline

CAS Number: 93-67-4
MDL Number: MFCD00025217
Molecular Formula: C12H10ClNO
Molecular Weight: 219.67
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 192
Melting point (°C): 43


Other Product Names / Synonyms: 5-Chloro-2-phenoxyaniline

Formula: C₁₂H₁₀ClNO
MW: 219.6700 g/mol
Boiling Pt: 192 °C
Melting Pt: 43 °C
MDL Number: MFCD00025217
CAS Number: 93-67-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a93bda1a149859ed514562ab2dd1f11489a5a9c&vid=6a93bda1a149859ed514562ab2dd1f11489a5a9c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR