1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)ethanone ≥98.0%

Supplier: TCI America

Synonyms: 3',4'-Ethylenedioxyacetophenone, 1,4-Benzodioxan-6-yl methyl ketone, 6-Acetyl-1,4-benzodioxane

A1629-5G
TCA1629-5GEA 63.8 USD
TCA1629-5G
1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)ethanone ≥98.0%
1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)ethanone

CAS Number: 2879-20-1
MDL Number: MFCD00006823
Molecular Formula: C10H10O3
Molecular Weight: 178.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 84


Other Product Names / Synonyms: 1,4-Benzodioxan-6-yl Methyl Ketone, 3',4'-Ethylenedioxyacetophenone

Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
Melting Pt: 84 °C
MDL Number: MFCD00006823
CAS Number: 2879-20-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3bb09d8acfb45dd49bb3cddd6b8dd5acb959ce79&vid=3bb09d8acfb45dd49bb3cddd6b8dd5acb959ce79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR