8-Amino-1-naphthalenesulfonic acid ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 8-Aminonaphthalene-1-sulphonic acid

A0350-100G
TCA0350-100GEA 74.3 USD
TCA0350-100G
8-Amino-1-naphthalenesulfonic acid ≥98.0% (by HPLC)
8-Amino-1-naphthalenesulphonic acid

CAS Number: 82-75-7
MDL Number: MFCD00035730
Molecular Formula: C10H9NO3S
Molecular Weight: 223.25
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: 1-Amino-8-naphthalenesulfonic Acid, 1-Naphthylamine-8-sulfonic Acid, Peri Acid

Formula: C₁₀H₉NO₃S
MW: 223.25 g/mol
MDL Number: MFCD00035730
CAS Number: 82-75-7
EINECS: 201-437-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48c398d65a74796e6be83169ea1713256f33c83e&vid=48c398d65a74796e6be83169ea1713256f33c83e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR