SideStep II Cell Lysis Analysis Kit, Agilent Technologies
Supplier: AGILENT TECH
400900
97066-404KT 272.24 USD
97066-404 ST400900
SideStep II Cell Lysis Analysis Kit, Agilent Technologies
Buffers Extraction Buffers
Progress from mammalian cells to qPCR or QRT-PCR experiments without nucleic acid purification.
  • The optimized cell lysis solution stabilizes RNA for at least 6 months
  • Samples are prepared ready for QRT-PCR analysis in minutes
  • The product ensures minimal RNA loss during preparation and analysis
  • The buffer employs a single-tube, solution-based format, rather than a column-based format
  • The process involves a single PBS wash followed by cell lysis in the SideStep lysis buffer
  • Immediately stabilize nucleic acid without further processing required, for more efficient analyses
The Agilent SideStep II Lysis and Stabilization Buffer uses a single-tube, solution-based format, rather than a column-based format. The patent-pending solution lyses cells in minutes and releases both RNA and DNA molecules into the solution. This immediately stabilizes nucleic acids without further processing required. The Cell Lysis and Protein Extraction Kit prevents sample loss and degradation, ensuring accurate gene quantification in downstream QRT-PCR. Large numbers of mammalian samples can be analyzed faster and more easily using the buffer, with minimal RNA loss. Skip nucleic acid purification steps in your applications today.
Our SideStep Lysis and Stabilization Buffer ensures accurate gene expression analysis by eliminating sample loss and degradation. The proprietary buffer lyses cells and immediately stabilizes the released nucleic acid for months. Several formats are available to enhance research versatility. Following a 10 minute room temperature lysis step, you can go directly from cells to qPCR amplification without stopping to purify. Advanced screening saves time and money by selecting the best RNA samples for purification. In addition, with a slight protocol modification the buffer can easily be used for detecting miRNA in cell lysates.
Precaución: For research use only. Not for use in diagnostic procedures.