Animal Whole Blood, Rockland Immunochemicals
Supplier: Rockland Immunochemical
R102-0100 R205-0100 R209-0100 R210-0100 R100-0050 R105-0050 R109-0100 R111-0100 R202-0100 R206-0050 R214-0100 R217-0100 R300-0050 R306-0050 R308-0050 R313-0100 R317-0100 R204-0100 R207-0100 R104-0100 R105-0100 R107-0050 R109-0050 R111-0050 R113-0100 R211-0100 R213-0100 R300-0100 R302-0050 R302-0100 R304-0100 R305-0050 R305-0100 R307-0100 R310-0050 R310-0100 R312-0050 R309-0050 R311-0100 R100-0100 R212-0050 R307-0050 R309-0100 R311-0050
RLR102EA 458.66 USD
RLR102 RLR205 RLR209 RLR210 RLR100-0050 RLR105-0050 RLR109-0100 RLR111-0100 RLR202 RLR206 RLR214 RLR217 RLR300-0050 RLR306-0050 RLR308-0050 RLR313 RLR317 RLR200 RLR204 RLR207 RLR104 RLR105-0100 RLR107-0050 RLR109-0050 RLR111-0050 RLR113 RLR211 RLR213 RLR300-0100 RLR302-0050 RLR302-0100 RLR304 RLR305-0050 RLR305-0100 RLR307-0100 RLR310-0050 RLR310-0100 RLR312 RLR309-0050 RLR311-0100 RLR100-0100 RLR212 RLR307-0050 RLR309-0100 RLR311-0050
Animal Whole Blood, Rockland Immunochemicals
Blood
Use for DNA isolation
Blood is obtained from healthy, non-medicated animals. All blood, except hamster, mouse, pigeon, and rat blood is sterile.
Información para pedidos: For additional coagulants, sizes, or species, contact your Avantor representative.
Order Now
SPECIFICATIONS
Environmentally Preferable