Tissue Powders for Adsorption, Rockland Immunochemicals
Supplier: Rockland Immunochemical
C101 C113 C120 C122 C103 C115 C116 C123 C124 C117 C119 C102 C121
RLC100EA 373.26 USD
RLC100 RLC101 RLC113 RLC120 RLC122 RLC103 RLC105 RLC106 RLC107 RLC109 RLC111 RLC114 RLC115 RLC116 RLC118 RLC123 RLC124 RLC108 RLC112 RLC117 RLC119 RLC102 RLC104 RLC110 RLC121
Tissue Powders for Adsorption, Rockland Immunochemicals
Media for Microbiology Dehydrated Media for Microbiology
Use for protocols that require a high nuclear protein content
Powders are produced by washing the various organs repeatedly in saline to remove blood, precipitated in acetone, and micronized to a fine powder. Keep container sealed and store at 2–8°C (36–46°F). All vials contain 1g of powder.