Anti-IgG Goat Polyclonal Antibody (DyLight® 405)

Supplier: Rockland Immunochemical
611-146-122
RL611-146-122EA 141 USD
RL611-146-122
Anti-IgG Goat Polyclonal Antibody (DyLight® 405)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 405
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR