Anti-IgG Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
609-1604
RL609-1604EA 258.06 USD
RL609-1604
Anti-IgG Goat Polyclonal Antibody (Biotin)
Antibodies
Secondary Goat Anti-IgG F(ab')2 Reacts with Human

Recommended Dilutions: ELISA: 1:20000 - 1:500,000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR