Anti-GAPDH Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
200-101-210-0100
RL200101210010EA 551.64 USD
RL200101210010
Anti-GAPDH Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: G3PD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Order Now

SPECIFICATIONS

Antigen Symbol GAPDH
Antigen Name Glyceraldehyde 3-Phosphate Dehydrogenase
Antigen Synonyms GAPDH|G3PD|CDABP0047|GAPD|Glyceraldehyde-3-phosphate dehydrogenase|Peptidyl-cysteine S-nitrosylase GAPDH|HEL-S-162eP|OK/SW-cl.12
Antibody Type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Rabbit
Host Goat
Concentration by UV absorbance at 280 nm
Storage Buffer 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR