Anti-ATM Mouse Polyclonal Antibody (Biotin) [clone: 10H11.E12]

Supplier: Rockland Immunochemical
200-306-400
RL200-306-400EA 657.16 USD
RL200-306-400
Anti-ATM Mouse Polyclonal Antibody (Biotin) [clone: 10H11.E12]
Antibodies
This antibody was tested by ELISA and western blotting against both the native and recombinant forms of the Protein. This reagent may also be suitable other biotin-streptavidin based assays.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 100 µl
Do not add Sodium Azide!

Type: Primary
Antigen: ATM
Clonality: Polyclonal
Clone: 10H11.E12
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen Symbol ATM
Antigen Name Ataxia telangiectasia mutated
Environmentally Preferable
Antigen Synonyms ATE|AT1|ATDC|ATD|TEL1|ATC|ATA|TELO1
Antibody Type Primary
Clonality Polyclonal
Conjugation Biotin
Clone 10H11.E12
Reactivity Human,Rat,Mouse
Host Mouse
Gene ID 472
Isotype IgG1 kappa
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
Modification phospho-Ser1981
Immunogen This antibody was produced from a synthetic peptide S-L-A-F-E-E-G-Sp-Q-S-T-T-I-S-S corresponding to aa 1974-1988 of human ATM.
Amino Acid Number 1974 to 1988
Sequence SLAFEEGSpQSTTISS
Format Antibody fraction
Form Lyophilized
Purification Protein A purified
Concentration 1.0 mg/ml by UV absorbance at 280 nm
Cross Adsorption No
Storage Buffer 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Gentamicin Sulfate. 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
Storage Temperature –20 °C
Shipping Temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR