Diskcover-47 Reservoir, Restek
Supplier: Restek
24029 24030
RK24029EA 147.29 USD
RK24029 RK24030
Diskcover-47 Reservoir, Restek
Reagent Reservoirs
The Diskcover-47 open-top reservoir allows users to pour up to 125 mL of sample directly onto the filter disk holder.
  • Easy installation on Diskcover-47 extraction disk holder