Capillary Installation Gauge for Bruker/Varian GCs for use with ¹/₁₆" ferrules, Restek
Supplier: Restek
22335
RK22335EA 98.84 USD
RK22335
Capillary Installation Gauge for Bruker/Varian GCs for use with ¹/₁₆" ferrules, Restek
Puller and Inserter Tools
Use a capillary installation gauge for Bruker/Varian GCs to seat ferrules onto a column for consistent installations.
  • For use with graphite ferrules only
Capillary installation gauges are for use with 1/16" ferrules.
Información para pedidos: For use with graphite ferrules only.
Order Now
SPECIFICATIONS
Environmentally Preferable