Replacement Ring Seal for MSD Conversion Fitting, Restek
Supplier: Restek
21313
RK21313PK 20.86 USD
RK21313
Replacement Ring Seal for MSD Conversion Fitting, Restek
Seals Sealing Rings
Flat, soft ring seal deforms and butt-seals against MSD interface.