Dual Vespel® Ring Inlet Seals for Agilent GCs, Restek
Supplier: Restek
21242 21243 21240 21245 21249 21241 21248 21239 21246 21238 21247 23418 23419 23420
RK21242PK 115.88 USD
RK21242 RK21243 RK21240 RK21245 RK21249 RK21241 RK21248 RK21239 RK21244 RK21246 RK21238 RK21247 10803-346 10856-132 10856-134
Dual Vespel® Ring Inlet Seals for Agilent GCs, Restek
Seals Mechanical Seals
Restek's patented dual Vespel® ring inlet seal greatly improves injection port performance—it stays sealed, even after repeated temperature cycles, without retightening the reducing nut.
  • Eliminate the need for a washer
  • Very little torque needed to make a leak-tight seal
  • Stainless steel seal for analyses of unreactive compounds
This seal features two soft Vespel® rings, one embedded in its top surface and the other embedded in its bottom surface. These rings eliminate the need for a washer, and ensure very little torque is needed to make a leak-tightMore Product Information