Inlet Liner Removal Tool, Restek
Supplier: Restek
20181
RK20181PK 55.4 USD
RK20181
Inlet Liner Removal Tool, Restek
Puller and Inserter Tools
With this tool, easily remove liners from injectors without any more burned fingers.
  • Made from high-temperature silicone
  • Won’t chip or crack the liner
Order Now
SPECIFICATIONS
Environmentally Preferable