2-Propanol 30% (v/v) in aqueous solution

Supplier: Ricca Chemical

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

R4207300-1A
RCR4207300-1AEA 70 USD
RCR4207300-1A
2-Propanol 30% (v/v) in aqueous solution
2-Propanol
Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: -89
Flash Pt: 12
MDL Number: MFCD00011674
CAS Number: 67-63-0
UN: 1987
ADR: 3 II
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 30 ± 1 % v/v


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR