Thallium ICP Standard, 1 ml = 1 mg Tl (1000 ppm Tl)Tl in 3% HNO₃, Ricca Chemical Company
Supplier: Ricca Chemical
RPTL1KN0-100N RPTL1KN0-500N
RCPTL1KN-100EA 134 USD
RCPTL1KN-100 RCPTL1KN-500
Thallium ICP Standard, 1 ml = 1 mg Tl (1000 ppm Tl)Tl in 3% HNO₃, Ricca Chemical Company
Standards Cationic Standards Thallium Standards