Sodium thiosulphate 0.100 (N/10) stabilised titrant ASTM D 1159

Supplier: Ricca Chemical
Warning

Synonyms: Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate

Stabilization: 0.02% Stabilised with sodium carbonate (Na₂CO₃)

7950-16
RC7950-16EA 28 USD
RC7950-16
Sodium thiosulphate 0.100 (N/10) stabilised titrant ASTM D 1159
Sodium thiosulphate

Sodium thiosulfate solution (0.1 N) ASTM (D 3869 A)
Sodium thiosulfate, 0.1 N APHA (4500-Cl B), standard
Sodium thiosulfate, 0.1 N TAPPI (T 624 cm-85)
Sodium thiosulfate, 0.1 N TAPPI (T 611 cm-97)
Sodium thiosulfate, 0.100 N APHA (4500-ClO2 E), standard
Sodium thiosulfate solution, 0.1 N APHA (4500-Cl C), standard
Sodium thiosulfate titrant, 0.1 N APHA (2350 E), standard
Sodium thiosulfate, 0.1 N APHA (2320 B)
Sodium thiosulfate, 0.1 M APHA (2310 B)
Sodium thiosulfate, 0.1 N TAPPI (T 254 cm-85)
Sodium thiosulfate, 0.1 N TAPPI (T 622 cm-84)
Sodium thiosulfate, 0.1 N volumetric solution ASTM (E 200)
Sodium thiosulfate, 0.1 N TAPPI (T 610 sp-97)
Sodium thiosulfate, standard solution (0.1 N) ASTM (D 3703)
Sodium thiosulfate, standard solution (0.1 N) ASTM (D 1832)
Sodium thiosulfate, standard solution (0.10 N) ASTM (D 1291)
Sodium thiosulfate, standard solution (0.1 M) ASTM (D 1159)
Sodium thiosulfate, 0.1 normal volumetric solution ACS (N/A)
Sodium thiosulphate, 0.1 M volumetric solution EP (3007300)
Sodium thiosulfate, 0.1 mol/L VS JP (N/A)
Sodium thiosulfate TS USP (N/A)
Sodium thiosulfate, 0.1 N volumetric solution USP (N/A)

Formula: Na₂S₂O₃
MW: 158.11 g/mol
Boiling Pt: 100
Melting Pt: 48
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
Merck Index: 12,08844

Order Now

Specification Test Results

Assay (vs. Potassium Iodate/Starch) 0.0999-0.1001 N at 20°C (NIST SRM 136)
Appearance Colorless liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR