Sodium hydroxide 10.0 N aqueous solution analytical reagent

Supplier: Ricca Chemical
Danger

Synonyms: Caustic soda

R7470000-1A
RC747032EA 114 USD
RC747032
Sodium hydroxide 10.0 N aqueous solution analytical reagent
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 10.00 ± 0.05 N at 20.0 °C
Heavy Metals (as Pb) max. 10 ppm
Iron max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR