Potassium permanganate 0.500 N (N/2) in aqueous solution

Supplier: Ricca Chemical

Synonyms: Permanganic acid potassium salt

R6410000-4C R6410000-1C
RC6410-1CS 1069 USD
RC6410-1 RC6410-32
Potassium permanganate 0.500 N (N/2) in aqueous solution
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 3082
ADR: 9 III
Merck Index: 12,07824

Order Now

Specification Test Results

Appearance Clear, purple, odorless
Assay at 20 °C (traceable to NIST SRM 40) 0.5000 ± 0.0002 N at 20.0 °C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR