Potassium iodide 0.100 M (0.100 N) aqueous solution Stabilised

Supplier: Ricca Chemical
Danger

Synonyms: Dipotassium diiodide, Kali iodide

6316-4
RC6316-4EA 155 USD
RC6316-4
Potassium iodide 0.100 M (0.100 N) aqueous solution Stabilised
Potassium iodide
Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330
Melting Pt: 686
Density: 1.0100 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
Merck Index: 13,07727

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Silver Nitrate/Eosin Y) 0.0998-0.1002 Molar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR