Nitrogen Standard, 1 ml = 0.1 mg N, 100 ppm N as Nitrate (442.7 ppm NO₃⁻), Ricca Chemical Company
Supplier: Ricca Chemical
R5457000-120A
RC5457-4EA 50 USD
RC5457-4
Nitrogen Standard, 1 ml = 0.1 mg N, 100 ppm N as Nitrate (442.7 ppm NO₃⁻), Ricca Chemical Company
Standards Anionic Standards Nitrate Standards