Mercury(II) nitrate 0.282 N

Supplier: Ricca Chemical

Synonyms: Mercury dinitrate, Mercuric nitrate

R4750000-500C R4750000-4C R4750000-1C
RC4750-16EA 116 USD
RC4750-16 RC47501 RC475032
Mercury(II) nitrate 0.282 N
Mercury(II) nitrate

1 ml = 10 mg Cl-

Formula: HgN₂O₆
MW: 324.6
Melting Pt: -38.9
Density: 4.3
MDL Number: MFCD00011038
CAS Number: 10045-94-0
UN: 1625
ADR: 6.1 II
Merck Index: 12,05933

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 0.2820 ± 0.0001 N at 20.0 °C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR