Crystal violet 1% (w/v) in glacial acetic acid indicator for nonaqueous titrations

Supplier: Ricca Chemical

Synonyms: CI 42555

R2483000-120C R2483000-500C
RC2483-4EA 66 USD
RC2483-4 RC2483-16
Crystal violet 1% (w/v) in glacial acetic acid indicator for nonaqueous titrations
Crystal violet
Formula: C₂₅H₃₀ClN₃
MW: 407.99 g/mol
MDL Number: MFCD00011750
CAS Number: 548-62-9
UN: 3077
ADR: 9 III
Merck Index: 13,04406

Order Now

Specification Test Results

Appearance Clear, violet, acetic odor
Suitability for Use Purple (Basic) - Blue-Green (Acidic)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR