Luzindole

Supplier: BACHEM AMERICAS INC

Synonyms: N-Acetyl-2-benzyl-tryptamine

4026095.0250 4026095.1000
Q-1885.0250BAEA 408.5 USD
Q-1885.0250BA Q-1885.1000BA
Luzindole
Luzindole

Sequence: N-Acetyl-2-benzyl-tryptamine


For luzindole see Q-1885.


Synonym(s): Luzindole

Formula: C₁₉H₂₀N₂O
MW: 292.382 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00672498
CAS Number: 117946-91-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR