Pierce™ TMTsixplex™ Protein Quantitation Kit and Label Reagent Sets, Thermo Scientific
Supplier: LIFE TECHNOLOGIES MS
90060 90061 90062 90063 90064 90065 90066 90068
PI90060EA 1026.46 USD
PI90060 PI90061 PI90062 PI90063 PI90064 PI90065 PI90066 PI90068
Pierce™ TMTsixplex™ Protein Quantitation Kit and Label Reagent Sets, Thermo Scientific
Protein Extraction Protein Extraction Kits
Pierce™ TMT™ Isobaric Mass Tagging kits and reagents enable multiplex quantitation of proteins extracted from cells and tissues using tandem mass spectrometry
  • Ideal for quantitative analysis of proteins for which no antibodies are available
  • Used in identification and quantitation of membrane and post-translationally modified proteins
Each mass-tagging reagent within a set has the same nominal mass (ie. isobaric) and chemical structure composed of an amine-reactive NHS-ester group, a spacer arm and an MS/MS reporter. The reagent sets can be used to label two or six peptide samples prepared from cells or tissues. For each sample, a unique reporter in the low mass region of the MS/MS spectrum (ie. 126-127 Da for TMTduplex and 126-131 Da for TMTsixplex Isobaric Label Reagents) is used to measure relative protein expression levels during peptide fragmentation.
The Tandem Mass Tag (TMT) reagent family consists of TMTduplex and TMTsixplex sets which are specially designed to enable a rapid and cost-effective transition from method development to high-throughput protein quantitation.
Información de suministro: Kits are supplied as standalone sets or in optimized kit formats containing all necessary reagents and controls for maximum flexibility.
Precaución: For Research Use Only. Not for use in diagnostic procedures.