Slide-A-Lyzer® MINI Dialysis Device Floats, Thermo Scientific
Supplier: Invitrogen
69588
PI69588PK 58.2 USD
PI69588
Slide-A-Lyzer® MINI Dialysis Device Floats, Thermo Scientific
Tubes Dialysis Tubes
The Slide-A-Lyzer® MINI Dialysis Device Floats are low-density polyethylene rafts designed to securely hold up to 25 Slide-A-Lyzer® MINI Dialysis Devices to permit array dialysis.
  • LDPE construction
  • Floating design
  • Not suitable for use at temperatures above 50 °C (122 °F)
Slide-A-Lyzer® MINI Dialysis Device Floats feature labeled rows (A to E) and columns (1 to 5) for easy tracking of individual samples. These floats are intended to hold Slide-A-Lyzer® MINI Dialysis Devices (0.1 ml size only), whose bottom surfaces are a dialysis interface composed ofMore Product Information