Pierce™ Slide-A-Lyzer® Dialysis Cassettes, Thermo Scientific
Supplier: Invitrogen
66003 66005 66012 66030 66107 66110 66130 66203 66205 66212 66230 66330 66332 66333 66335 66370 66372 66373 66380 66381 66382 66383 66384 66385 66453 66454 66455 66456 66707 66710 66807 66810 66811 66830
PI66003EA 161.7 USD
PI66003 PI66005 PI66012 PI66030 PI66107 PI66110 PI66130 PI66203 PI66205 PI66212 PI66230 PI66330 PI66332 PI66333 PI66335 PI66370 PI66372 PI66373 PI66380 PI66381 PI66382 PI66383 PI66384 PI66385 PI66453 PI66454 PI66455 PI66456 PI66707 PI66710 PI66807 PI66810 PI66811 PI66830
Pierce™ Slide-A-Lyzer® Dialysis Cassettes, Thermo Scientific
Tubing Dialysis Tubing
For sample dialysis, low molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and concentration. This type of membrane is compatible with a number of common chemicals and buffers. The colour-coded transparent frames of the Slide-A-Lyzer® cassette allow instant recognition of the MWCO of the membrane and make viewing the needle easier during sample injection.
  • Quick and convenient
  • Durable and high recovery
  • Plastic construction
Require approximately half the time of traditional dialysis tubing and produce sample volumeMore Product Information