Pierce™ Streptavidin Plus UltraLink™ Resin, Thermo Scientific
Supplier: LIFE TECHNOLOGIES MS
53116 53117
PI53116EA 606.77 USD
PI53116 PI53117
Pierce™ Streptavidin Plus UltraLink™ Resin, Thermo Scientific
Protein Purification Protein Purification Resins
Pierce™ Streptavidin Plus UltraLink™ resin consists of purified recombinant streptavidin that has been covalently immobilized onto a beaded acrylamide support
  • Acrylamide based support provides an alternative to crosslinked beaded agarose supports for medium pressure applications
  • Streptavidin Plus UltraLink™ resin binding capacity: >4 mg of biotinylated BSA/mL of resin
UltraLink™ beads are hydrophilic, charge free, highly crosslinked, rigid and durable which makes the affinity resin suitable for a variety of batch or column type affinity procedures that require medium pressure chromatography.
Precaución: For Research Use Only. Not for use in diagnostic procedures.