Water LC-MS, Pierce™

Supplier: Thermo Fisher Scientific

Synonyms: Hydrogen oxide

T51140-K2
PI51140EA 99.78 USD
PI51140
Water LC-MS, Pierce™
Water

Thermo Scientific Pierce Water, LC-MS Grade is an ultrapure, LC-MS grade preparation with low UV-absorptivity that makes it suitable and reliable for use in HPLC and mass spectrometry applications.


  • Purity and quality validated with 30 tests for use in LC-MS applications
  • 0.2 micron filtered, packed in solvent-rinsed amber glass bottles with Teflon TFE-lined fluorocarbon caps for ultimate protection
  • Sealed under nitrogen atmosphere to eliminate absorption of unknown atmospheric gases
  • High lot-to-lot consistency and reliability


Low UV absorptivity makes this product ideal for use in HPLC applications.

Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.


Precaución: For Research Use Only. Not for use in diagnostic procedures.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR