Pierce™ Fab Micro Preparation Kit, Thermo Scientific
Supplier: LIFE TECHNOLOGIES MS
44685 44985
PI44685EA 745.38 USD
PI44685 PI44985
Pierce™ Fab Micro Preparation Kit, Thermo Scientific
Protein Purification Protein Purification Kits
Thermo Scientific Pierce Fab Micro Preparation Kit uses immobilized papain protease to digest human or mouse IgG antibodies to make separate Fab and Fc fragments and subsequently to purify the Fab using Protein A agarose.
  • High capacity—use the micro kit for 25 to 250 µg IgG.
  • Enzyme-free digestion products—Immobilized Papain (beaded agarose resin) provides for control of the digestion reaction and complete removal of resulting antibody fragments from the proteolytic enzyme
  • Digestion and final recovery of purified Fab fragments occurs in neutral pH sodium phosphate buffer, suitable for storage or immediate use in typical applications
  • Spin format reduces sample processing time
  • Protocols are included for multiple species and IgG subclasses, as well as sample size and concentration
Suitable for human and other species of IgG. The kit procedure is optimized for human, mouse and rabbit IgG. Papain-based digestion is effective for certain other species and subclasses of IgG (e.g., goat and pig), although the purification step requires that the antibody effectively binds to Protein A.
Información de suministro: Each standard kit includes immobilized papain, 1.25 mL; cysteine-HCl, 1 mg; Fab digestion buffer, 120 mL; NAb™ Protein A Plus spin columns, 1 x 1 mL; BupH™ PBS, 2 packs (makes 1L); IgG elution buffer, 120 mL; spin columns, 10 x 0.8 mL; microcentrifuge tubes, 30 x 2 mL; and Zeba™ desalting columns, 7K MWCO, 10 x 2 mL. Each micro kit includes immobilized papain, 0.5 mL; cysteine-HCl, 1 mg; Fab digestion buffer, 55 mL; NAb™ Protein A Plus spin columns, 2 x 0.2 mL; BupH™ PBS, 2 packs (makes 1L); IgG elution buffer, 50 mL; spin columns, 10 x 0.8 mL; microcentrifuge tubes, 30 x 2 mL; and Zeba™ desalting columns, 7K MWCO, 10 x 0.5 mL.
Precaución: For Research Use Only. Not for use in diagnostic procedures. Store at 4–8˚C.